Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,3-dihydrobenzofuran 98%

Supplier: AMBEED, INC
Warning

A358371-10G A358371-1G A358371-5G
76694-178EA 32 USD InStock
76694-178 76694-180 77626-692
5-Bromo-2,3-dihydrobenzofuran 98%
5-Bromo-2,3-dihydrobenzofuran
Precaución: for Research Use Only


Formula: C₈H₇BrO
MW: 199.05 g/mol
MDL Number: MFCD02677716
CAS Number: 66826-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty