Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Hydroxycoumarin-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A358382-100MG A358382-1G A358382-250MG A358382-5G A358382-25G
76694-182EA 12 USD InStock
76694-182 76694-184 76694-186 77601-588 77626-694
7-Hydroxycoumarin-3-carboxylic acid 98%
7-Hydroxycoumarin-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₆O₅
MDL Number: MFCD00017491
CAS Number: 779-27-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty