Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-4-isopropylaniline 98%

Supplier: AMBEED, INC
Warning

A368264-100G A368264-1G A368264-5G
76694-292EA 179 USD InStock
76694-292 76694-294 76694-296
2-Bromo-4-isopropylaniline 98%
2-Bromo-4-isopropylaniline
Precaución: for Research Use Only


Formula: C₉H₁₂BrN
MW: 214.1
Density: 1.35
MDL Number: MFCD00051585
CAS Number: 51605-97-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty