Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Benzyloxycarbonylamino)glutarimide 97%

Supplier: AMBEED, INC
Warning

A377646-100MG A377646-1G A377646-250MG
76694-414EA 22 USD InStock
76694-414 76694-416 76694-418
2-(Benzyloxycarbonylamino)glutarimide 97%
2-(Benzyloxycarbonylamino)glutarimide
Precaución: for Research Use Only


Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD01075195
CAS Number: 24666-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty