Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Delavirdine mesylate 99+%

Supplier: AMBEED, INC
Danger

A380147-100MG A380147-10MG A380147-1MG A380147-50MG
76694-458EA 119 USD InStock
76694-458 76694-460 76694-462 76694-464
Delavirdine mesylate 99+%
Delavirdine mesylate
Precaución: for Research Use Only


Formula: C₂₃H₃₂N₆O₆S₂
MDL Number: MFCD00866948
CAS Number: 147221-93-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty