Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) 98%

Supplier: AMBEED, INC
Warning

A380529-1G A380529-250MG A380529-25G A380529-5G A380529-100G
76694-466EA 26 USD InStock
76694-466 76694-468 76694-470 76694-472 77626-744
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) 98%
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Precaución: for Research Use Only


Formula: C₅₅H₄₅ClOP₃Ru
MW: 951.405 g/mol
MDL Number: MFCD00049804
CAS Number: 16971-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty