Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(I) bromide 99.5%

Supplier: AMBEED, INC
Danger

A381261-100G A381261-25G
76694-480EA 20 USD InStock
76694-480 76694-482
Copper(I) bromide 99.5%
Copper(I) bromide
Precaución: For research use only.


Formula: BrCu
MW: 143.45
Boiling Pt: 1345
Melting Pt: 504
Density: 4.71
MDL Number: MFCD00010969
CAS Number: 7787-70-4
UN: 3077
ADR: 9 Ⅲ
Merck Index: 13,02689
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty