Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butylamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A383644-100G A383644-1G A383644-25G A383644-5G
76694-520EA 54 USD InStock
76694-520 76694-522 76694-524 76694-526
Butylamine hydrochloride 98%
Butylamine hydrochloride


Formula: C₄H₁₁N
MDL Number: MFCD00043791
CAS Number: 3858-78-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty