Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium benzenesulfonate 95% (contains < 15% H₂O)

Supplier: AMBEED, INC
Warning

A383717-100G A383717-25G A383717-500G
76694-528EA 7 USD InStock
76694-528 76694-530 76694-532
Sodium benzenesulfonate 95% (contains < 15% H₂O)
Sodium benzenesulfonate
Precaución: for Research Use Only


Formula: C₆H₆O₃S
MW: 180.15 g/mol
MDL Number: MFCD00065179
CAS Number: 515-42-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty