Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Pyridyl)acrylic acid 95%

Supplier: AMBEED, INC
Warning

A385192-100G A385192-10G A385192-1G A385192-25G A385192-5G
76694-546EA 143 USD InStock
76694-546 76694-548 76694-550 76694-552 76694-554
3-(3-Pyridyl)acrylic acid 95%
3-(3-Pyridyl)acrylic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₂
MDL Number: MFCD00006410
CAS Number: 1126-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty