Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyl acrylate 97% stabilised

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A386639-100G A386639-10G A386639-1G A386639-25G A386639-5G
76694-582EA 22 USD InStock
76694-582 76694-584 76694-586 76694-588 76694-590
Benzyl acrylate 97% stabilised
Benzyl acrylate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00048147
CAS Number: 2495-35-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty