Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane) 98%

Supplier: AMBEED, INC
Warning

A390373-100MG A390373-250MG A390373-1G
76694-636EA 12 USD InStock
76694-636 76694-638 77626-778
Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane) 98%
Benzo-12-crown-4 (Benzo-1,4,7,10-tetraoxacyclododecane)
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₄
MDL Number: MFCD00059934
CAS Number: 14174-08-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty