Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benfotiamine 97%

Supplier: AMBEED, INC
Warning

A390519-250MG A390519-1G A390519-25G A390519-5G
76694-650EA 5 USD InStock
76694-650 77626-780 77626-782 77626-784
Benfotiamine 97%
Benfotiamine
Precaución: for Research Use Only


Formula: C₁₉H₂₃N₄O₆PS
MDL Number: MFCD00057343
CAS Number: 22457-89-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty