Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3-Dimethylbenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A391680-100G A391680-10G A391680-1G A391680-25G A391680-5G
76694-676EA 171 USD InStock
76694-676 76694-678 76694-680 76694-682 76694-684
2,3-Dimethylbenzaldehyde 95%
2,3-Dimethylbenzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₀O
MDL Number: MFCD00798004
CAS Number: 5779-93-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty