Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 7-methyl-1H-indole-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A393510-100MG A393510-1G A393510-250MG A393510-5G
76694-700EA 7 USD InStock
76694-700 76694-702 76694-704 76694-706
Methyl 7-methyl-1H-indole-3-carboxylate 95%
Methyl 7-methyl-1H-indole-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MDL Number: MFCD06204262
CAS Number: 773134-49-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty