Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(tert-Butoxycarbonyl)-L-methioninol 97%

Supplier: AMBEED, INC
Danger

A410832-10G A410832-1G A410832-25G A410832-5G
76694-832EA 55 USD InStock
76694-832 76694-834 76694-836 76694-838
N-(tert-Butoxycarbonyl)-L-methioninol 97%
N-(tert-Butoxycarbonyl)-L-methioninol
Precaución: for Research Use Only


Formula: C₁₀H₂₁NO₃S
MW: 235.34 g/mol
MDL Number: MFCD00235939
CAS Number: 51372-93-1
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty