Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Hydroxyethyl)indole 98%

Supplier: AMBEED, INC
Warning

A434396-100MG A434396-1G A434396-25G A434396-50MG A434396-5G
76695-092EA 6 USD InStock
76695-092 76695-094 76695-096 76695-098 76695-100
3-(2-Hydroxyethyl)indole 98%
3-(2-Hydroxyethyl)indole
Precaución: for Research Use Only


Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 358
MDL Number: MFCD00005659
CAS Number: 526-55-6
Merck Index: 12,09930
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty