Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-5438 99%

Supplier: AMBEED, INC
Warning

A457556-100MG A457556-10MG A457556-1MG A457556-50MG A457556-5MG
76695-292EA 472 USD InStock
76695-292 76695-294 76695-296 76695-298 76695-300
AZD-5438 99%
AZD-5438
Precaución: for Research Use Only


Formula: C₁₈H₂₁N₅OS
MW: 371.46 g/mol
MDL Number: MFCD11112135
CAS Number: 602306-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty