Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[cd]indol-2(1H)-one 95%

Supplier: AMBEED, INC
Warning

A466724-1000G A466724-500G A466724-100G
76695-412EA 143 USD InStock
76695-412 76695-414 76695-416 77572-470
Benzo[cd]indol-2(1H)-one 95%
Benzo[cd]indol-2(1H)-one
Precaución: for Research Use Only


Formula: C₁₁H₇NO
MW: 169.18 g/mol
MDL Number: MFCD00009748
CAS Number: 130-00-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty