Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(carboxymethyl)trithiocarbonate 98%

Supplier: AMBEED, INC
Warning

A468103-100G A468103-25G A468103-5G
76695-424EA 159 USD InStock
76695-424 77594-478 77602-044
Bis(carboxymethyl)trithiocarbonate 98%
Bis(carboxymethyl)trithiocarbonate
Precaución: for Research Use Only


Formula: C₅H₆O₄S₃
MDL Number: MFCD00004357
CAS Number: 6326-83-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty