Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluorobenzo[b]thiophene-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A475370-1G A475370-250MG
76695-446EA 50.93 USD InStock
76695-446 76695-448 76695-450
4-Fluorobenzo[b]thiophene-2-carboxylic acid 95%
4-Fluorobenzo[b]thiophene-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₅FO₂S
MDL Number: MFCD03425782
CAS Number: 310466-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty