Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1H-Imidazol-1-yl)phenol 95%

Supplier: AMBEED, INC
Warning

A478357-100MG A478357-1G A478357-25G A478357-5G
76695-490EA 5 USD InStock
76695-490 76695-492 76695-494 76695-496
4-(1H-Imidazol-1-yl)phenol 95%
4-(1H-Imidazol-1-yl)phenol
Precaución: for Research Use Only


Formula: C₉H₈N₂O
MDL Number: MFCD00005281
CAS Number: 10041-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty