Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Angelic acid 98%

Supplier: AMBEED, INC
Danger

A478510-1MG A478510-100MG A478510-250MG
76695-498EA 5 USD InStock
76695-498 77626-950 77626-952
Angelic acid 98%
Angelic acid
Precaución: for Research Use Only


Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00002654
CAS Number: 565-63-9
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty