Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-acetylbenzoate 98%

Supplier: AMBEED, INC
Warning

A500909-100G A500909-10G A500909-1G A500909-25G A500909-500G A500909-5G
76695-698EA 102 USD InStock
76695-698 76695-700 76695-702 76695-704 76695-706 76695-708
Methyl-4-acetylbenzoate 98%
Methyl 4-acetylbenzoate
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
MDL Number: MFCD00216474
CAS Number: 3609-53-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty