Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ammonium hexachloropalladate(IV) 98%

Supplier: AMBEED, INC
Warning

A502339-1G A502339-250MG
76695-712EA 67 USD InStock
76695-712 76695-714
Ammonium hexachloropalladate(IV) 98%
Ammonium hexachloropalladate(IV)
Precaución: for Research Use Only


Formula: H₈Cl₆N₂Pd
MW: 355.21
Density: 2.418
MDL Number: MFCD00015959
CAS Number: 19168-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty