Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyloxyacetaldehyde dimethyl acetal 95%

Supplier: AMBEED, INC
Warning

A504182-100G A504182-10G A504182-25G A504182-5G
76695-730EA 302 USD InStock
76695-730 76695-732 76695-734 76695-736
Benzyloxyacetaldehyde dimethyl acetal 95%
Benzyloxyacetaldehyde dimethyl acetal
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₃
MW: 196.24 g/mol
MDL Number: MFCD00082233
CAS Number: 127657-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty