Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2-Tris(tert-butoxycarbonyl)hydrazine 97%

Supplier: AMBEED, INC
Warning

A533881-1G A533881-5G
76695-970EA 23 USD InStock
76695-970 77627-086
1,1,2-Tris(tert-butoxycarbonyl)hydrazine 97%
1,1,2-Tris(tert-butoxycarbonyl)hydrazine
Precaución: For research use only.


Formula: C₁₅H₂₈N₂O₆
MW: 332.39 g/mol
MDL Number: MFCD04973130
CAS Number: 185456-26-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty