Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Glu-OtBu 98%

Supplier: AMBEED, INC
Warning

A539613-100G A539613-10G A539613-1G A539613-25G A539613-5G
76696-026EA 231 USD InStock
76696-026 76696-028 76696-030 76696-032 76696-034
Boc-D-Glu-OtBu 98%
Boc-D-Glu-OtBu
Precaución: for Research Use Only


Formula: C₁₄H₂₅NO₆
MW: 303.355 g/mol
MDL Number: MFCD00076926
CAS Number: 73872-71-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty