Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

copper(ii)hexafluor-2,4-pentanedionate 98%

Supplier: AMBEED, INC
Warning

A540069-1G A540069-250MG A540069-5G
76696-042EA 16 USD InStock
76696-042 76696-044 76696-046
copper(ii)hexafluor-2,4-pentanedionate 98%
copper(ii)hexafluor-2,4-pentanedionate
Precaución: for Research Use Only


Formula: C₁₀H₂CuF₁₂O₄
MW: 477.64 g/mol
MDL Number: MFCD00064754
CAS Number: 14781-45-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty