Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyldimethylhexadecylammonium chloride 98% (contains < 10% H₂O)

Supplier: AMBEED, INC
Danger

A545178-10G A545178-25G
76696-090EA 5 USD InStock
76696-090 76843-850
Benzyldimethylhexadecylammonium chloride 98% (contains < 10% H₂O)
Benzyldimethylhexadecylammonium chloride
Precaución: for Research Use Only


Formula: C₂₅H₄₆ClN
MW: 396.10 g/mol
MDL Number: MFCD00011625
CAS Number: 122-18-9
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty