Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Cyanophenyl)piperazine 98%

Supplier: AMBEED, INC
Warning

A546893-1G A546893-250MG A546893-5G
76696-092EA 42 USD InStock
76696-092 76696-094 76696-096
1-(3-Cyanophenyl)piperazine 98%
1-(3-Cyanophenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₁H₁₃N₃
MDL Number: MFCD06796181
CAS Number: 178928-58-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty