Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Piperazin-1-ylnicotinonitrile 95%

Supplier: AMBEED, INC
Danger

A547190-250MG A547190-5G
76696-098EA 12 USD InStock
76696-098 76696-100
6-Piperazin-1-ylnicotinonitrile 95%
6-Piperazin-1-ylnicotinonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₂N₄
MDL Number: MFCD00833024
CAS Number: 149554-29-0
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty