Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,4-Trifluoro-1-phenyl-1,3-butanedione 97%

Supplier: AMBEED, INC
Warning

A551229-100G A551229-10G A551229-1G A551229-25G A551229-5G
76696-130EA 58 USD InStock
76696-130 76696-132 76696-134 76696-136 76696-138
4,4,4-Trifluoro-1-phenyl-1,3-butanedione 97%
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Precaución: for Research Use Only


Formula: C₁₀H₇F₃O₂
MDL Number: MFCD00000425
CAS Number: 326-06-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty