Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Bromo-cAMP sodium salt 98%

Supplier: AMBEED, INC
Warning

A551904-100MG A551904-10MG A551904-1MG A551904-25MG A551904-50MG
76696-148EA 275 USD InStock
76696-148 76696-150 76696-152 76696-154 76696-156
8-Bromo-cAMP sodium salt 98%
8-Bromo-cAMP sodium salt
Precaución: for Research Use Only


Formula: C₁₀H₁₀BrN₅NaO₆P
MW: 430.08 g/mol
MDL Number: MFCD00005844
CAS Number: 76939-46-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty