Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzylbis(triphenylphosphine)palladium(II) chloride 98%

Supplier: AMBEED, INC
Warning

A556630-100MG A556630-1G A556630-250MG
76696-176EA 72 USD InStock
76696-176 76696-178 76696-180
Benzylbis(triphenylphosphine)palladium(II) chloride 98%
Benzylbis(triphenylphosphine)palladium(II) chloride
Precaución: for Research Use Only


Formula: C₄₃H₃₇ClP₂Pd
MW: 757.59 g/mol
MDL Number: MFCD00010395
CAS Number: 22784-59-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty