Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-5-phenylhydantoin 98%

Supplier: AMBEED, INC
Warning

A559945-100G A559945-1G A559945-25G A559945-5G
76696-218EA 417 USD InStock
76696-218 76696-220 76696-222 76696-224
5-Methyl-5-phenylhydantoin 98%
5-Methyl-5-phenylhydantoin
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂O₂
MDL Number: MFCD00005265
CAS Number: 6843-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty