Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-1,2,4,5-tetramethylbenzene 95%

Supplier: AMBEED, INC
Warning

A563968-100G A563968-1G A563968-25G A563968-5G
76696-240EA 482 USD InStock
76696-240 76696-242 76696-244 76696-246
3-Bromo-1,2,4,5-tetramethylbenzene 95%
3-Bromo-1,2,4,5-tetramethylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₃Br
MDL Number: MFCD00000070
CAS Number: 1646-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty