Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N-Diethyl-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine 98%

Supplier: AMBEED, INC
Warning

A569374-10MG A569374-50MG A569374-5MG A569374-1MG
76696-278EA 8 USD InStock
76696-278 76696-280 76696-282 77627-178
N,N-Diethyl-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine 98%
N,N-Diethyl-5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₅
MDL Number: MFCD00193104
CAS Number: 15421-84-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty