Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N,N,2-Trimethyl-5-nitrobenzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A575103-100MG A575103-10MG A575103-25MG A575103-50MG A575103-5MG A575103-250MG
76696-344EA 106 USD InStock
76696-344 76696-346 76696-348 76696-350 76696-352 77627-192
N,N,2-Trimethyl-5-nitrobenzenesulfonamide 98%
N,N,2-Trimethyl-5-nitrobenzenesulfonamide
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₄S
MW: 244.27 g/mol
MDL Number: MFCD03039912
CAS Number: 433695-36-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty