Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Dehydrocholesterol 95%

Supplier: AMBEED, INC
Warning

A584741-1G A584741-25G A584741-5G A584741-100G
76696-414EA 13 USD InStock
76696-414 76696-416 76696-418 77627-208
7-Dehydrocholesterol 95%
7-Dehydrocholesterol
Precaución: for Research Use Only


Formula: C₂₇H₄₄O
MDL Number: MFCD00003624
CAS Number: 434-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty