Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Butyl-10-undecenoate 95+%

Supplier: AMBEED, INC
Warning

A586085-100G A586085-25G
76696-426EA 49 USD InStock
76696-426 76696-428
Butyl-10-undecenoate 95+%
Butyl-10-undecenoate
Precaución: for Research Use Only


Formula: C₁₅H₂₈O₂
MDL Number: MFCD00048776
CAS Number: 109-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty