Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate 98%

Supplier: AMBEED, INC
Warning

A588076-10G A588076-1G A588076-5G
76696-438EA 1290 USD InStock
76696-438 76696-440 76696-442
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate 98%
Bis(cyclopentadienyl)cobalt(III) hexafluorophosphate
Precaución: for Research Use Only


Formula: C₁₀H₁₀CoF₆P
MDL Number: MFCD00011874
CAS Number: 12427-42-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty