Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-4-Decen-1-ol 90+%

Supplier: AMBEED, INC
Warning

A594932-100G A594932-1G A594932-25G A594932-5G
76696-466EA 680 USD InStock
76696-466 76696-468 76696-470 76696-472
cis-4-Decen-1-ol 90+%
cis-4-Decen-1-ol
Precaución: for Research Use Only


Formula: C₁₀H₂₀O
MDL Number: MFCD00015568
CAS Number: 57074-37-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty