Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A601281-100MG A601281-1G A601281-250MG A601281-5G
76696-520EA 13 USD InStock
76696-520 76696-522 76696-524 76696-526
7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid 98%
7-Nitro-2,3-dihydro-1,4-benzodioxine-6-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₇NO₆
MDL Number: MFCD01116988
CAS Number: 57672-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty