Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-1-methoxy-3-nitrobenzene 98%

Supplier: AMBEED, INC
Warning

A604111-100MG A604111-10G A604111-1G A604111-250MG A604111-25G
76696-550EA 5 USD InStock
76696-550 76696-552 76696-554 76696-556 76696-558
2-Chloro-1-methoxy-3-nitrobenzene 98%
2-Chloro-1-methoxy-3-nitrobenzene
Precaución: for Research Use Only


Formula: C₇H₆ClNO₃
MDL Number: MFCD11035883
CAS Number: 3970-39-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty