Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-Tyr-OBZL 98%

Supplier: AMBEED, INC
Warning

A607451-100G A607451-1G A607451-25G A607451-5G
76696-568EA 260 USD InStock
76696-568 76696-570 76696-572 76696-574
Boc-Tyr-OBZL 98%
Boc-Tyr-OBZL
Precaución: for Research Use Only


Formula: C₂₁H₂₅NO₅
MW: 371.433 g/mol
MDL Number: MFCD00190840
CAS Number: 19391-35-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty