Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bzl-L-Ala-OMe·HCI 98%

Supplier: AMBEED, INC
Warning

A618405-100G A618405-25G A618405-5G
76696-676EA 339 USD InStock
76696-676 76696-678 76696-680
Bzl-L-Ala-OMe·HCI 98%
Bzl-L-Ala-OMe·HCI
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂·HCl
MW: 229.74 g/mol
MDL Number: MFCD02684375
CAS Number: 19460-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty