Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Methyl-3-(trifluoromethyl)benzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A618527-1G A618527-25G A618527-5G
76696-682EA 24 USD InStock
76696-682 76696-684 77627-252
α-Methyl-3-(trifluoromethyl)benzyl alcohol 97%
alpha-Methyl-3-(trifluoromethyl)benzyl alcohol
Precaución: for Research Use Only


Formula: C₉H₉F₃O
MDL Number: MFCD00021868
CAS Number: 454-91-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty