Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Acridinecarboxylic acid hydrate 97%

Supplier: AMBEED, INC
Warning

A622428-1G A622428-5G
76696-716EA 12 USD InStock
76696-716 76696-718
9-Acridinecarboxylic acid hydrate 97%
9-Acridinecarboxylic acid hydrate
Precaución: for Research Use Only


Formula: C₁₄H₁₁NO₃
MW: 241.24 g/mol
MDL Number: MFCD00149578
CAS Number: 332927-03-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty