Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Hydroxyquinoline-5-sulfonic acid monohydrate 98%

Supplier: AMBEED, INC
Warning

A623815-100G A623815-10G A623815-25G A623815-500G A623815-5G
76696-738EA 65 USD InStock
76696-738 76696-740 76696-742 76696-744 76696-746
8-Hydroxyquinoline-5-sulfonic acid monohydrate 98%
8-Hydroxyquinoline-5-sulphonic acid monohydrate
Precaución: for Research Use Only


Formula: C₉H₇NO₄S·1H₂O
MDL Number: MFCD00149482
CAS Number: 283158-18-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty